Orders LinkContactUsLinkComponent
Test tube baskets
Test tube baskets
127×102×102 mm
Codice catalogo 489-2210
Nome del fornitore:   Bel-Art Products, a Part of SP
Test tube baskets
Codice catalogo 489-2210
Nome del fornitore:  Bel-Art Products, a Part of SP
Test tube baskets
Test tube baskets
127×102×102 mm
Price
SFr. 77.50
Log in per disponibilità
1 * 1 ST
Product Details & Documents
In PP.
  • Very good chemical resistance
  • Very good temperature resistance
  • Sterilisable and autoclavable
These test tube baskets are designed for rinsing, transporting and storing laboratory glassware.
Colour: White
Specifiche
  • L×P×A:
    127×102×102 mm