Burner, Teclu
Teclu burner with air regulation needle valve, natural gas, tubing Ø 14 mm, Gas naturale, 17 mm
Codice catalogo 451-1341
Nome del fornitore: USBECK Laborgeräte
Vai a
Product Details & Documents
Nickel plated brass base, in powder coated grey.
- With air regulation valve and throttle screw
- Without pilot light
Height: 165 mm
Specifiche
- Descrizione:Teclu burner with air regulation needle valve, natural gas, tubing Ø 14 mm
- Ø testa:17 mm
- Per:Gas naturale