Adhesive seals
Pellicola adesiva EASYseal, non sterile, 80×140 mm, Transparent, -
Catalog # 736-0127
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
These adhesive seals offer an alternative to heat seals or sealing mats. EASYseal™ is a transparent adhesive film designed to cover microplates to protect samples from evaporation and contamination. It is suitable for routine laboratory use, including ELISA applications. BREATHseal™ is a gas permeable, pierceable membrane coated with an acrylic adhesive. It is ideal for bacterial, yeast and cell culture applications, assuring the protection of cell contents from contamination whilst maintaining oxygenation.
- Free from RNase, DNase and human DNA
- Non pyrogenic
Specifications
- Proprietà ottica:Transparent
- Sterili:-
- Descrizione:Pellicola adesiva EASYseal, non sterile, 80×140 mm
- Seq:02