Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Powder spatulas, micro

Powder spatulas, micro

140 mm, 3 mm
Codice catalogo 231-2150
Nome del fornitore:   HAMMACHER KARL
Powder spatulas, micro
Codice catalogo 231-2150
Nome del fornitore:  HAMMACHER KARL
Powder spatulas, micro
Powder spatulas, micro

140 mm, 3 mm

Price
SFr. 41.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Single-ended powder spatulas made from WIRONIT steel.

Specifiche
  • Lunghezza:
    140 mm
  • Seq:
    01
  • Larghezza lama:
    3 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.