Funnels with short stem
38 mm, 11 mm, 100 mm
Codice catalogo 221-1155
Nome del fornitore: SEMADENI
Vai a
Product Details & Documents
Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
- Easy to clean
- Good chemical resistance
- Good temperature resistance
Specifiche
- Ø esterno imbuto:100 mm
- Ø esterno stelo:11 mm
- Lunghezza asta:38 mm



