Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

70 mm, 18 mm, 180 mm
Codice catalogo 221-1152
Nome del fornitore:   SEMADENI
Funnels with short stem
Codice catalogo 221-1152
Nome del fornitore:  SEMADENI
Funnels with short stem
Funnels with short stem

70 mm, 18 mm, 180 mm

Price
SFr. 6.60
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.

  • Easy to clean
  • Good chemical resistance
  • Good temperature resistance
Specifiche
  • Ø esterno imbuto:
    180 mm
  • Ø esterno stelo:
    18 mm
  • Lunghezza asta:
    70 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.