Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Funnels, with short stem, PP

Funnels, with short stem, PP

80 mm, 11 mm, 100 mm
Catalog # 221-1150
Supplier:   SEMADENI
Funnels, with short stem, PP
Catalog # 221-1150
Supplier:  SEMADENI
Funnels, with short stem, PP
Funnels, with short stem, PP

80 mm, 11 mm, 100 mm

Price
SFr. 7.50
Log in for availability
1 * 1 ST
Product Details & Documents

Funnels, made of PP, translucent, with slim stem.

  • Good chemical resistance
  • Good temperature resistance
  • Autoclavable
Specifications
  • Ø esterno imbuto:
    100 mm
  • Ø esterno stelo:
    11 mm
  • Lunghezza asta:
    80 mm