Wire basket, round
200×200 mm
Nome del fornitore: DELTALAB
Vai a
Product Details & Documents
Stainless steel.
These round baskets feature a folding handle for more convenience. The base of these baskets is reinforced by a thick mesh for added strength.
Informazioni per l'ordine: Models with inner crossed dividers are available.
Documents
Certificates
Cerca per numero di lottoSpecifiche
- Ø×A:200×200 mm
- Seq:02