Instrument dishes
Coperchio per capsule 216-1404, 220x150x10 mm
Codice catalogo 216-1414
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Thick-walled glass.
- Can be sterilised in hot air up to 180 °C
- Robust
- Stainless steel lids are available separately
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Specifiche
- Descrizione:Coperchio per capsule 216-1404
- Seq:11
- L×P×A:220x150x10 mm