Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
PCR manual workstation

PCR manual workstation

PCR rack with lid, yellow
Codice catalogo 212-9156
Nome del fornitore:   DELTALAB
PCR manual workstation
Codice catalogo 212-9156
Nome del fornitore:  DELTALAB
PCR manual workstation
PCR manual workstation

PCR rack with lid, yellow

Price
129.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents

This storage/working rack, with lid, is designed to work with different PCR volumes at the same time. It can accommodate up to 32 individual 0,2 ml PCR tubes, 24× 1,5/2,0 ml and 16× 0,5 ml microtubes.

  • Tube wells are alphanumerically matrixed with raised bold type for accurate sample identification
  • Rack has a transparent lid for easy viewing of samples
  • Frosted writing panels on four sides allow labelling or writing
  • Supplied with a removable rack accommodating 0,2 ml PCR tubes

Main rack W×D×H: 215×118×50 mm
Removable rack W×D×H: 125×85×15 mm

Documents

Specifiche
  • Descrizione:
    PCR rack with lid, yellow
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.