Blu Timolo ACS
Bottiglia di plastica, 2 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 76-61-9 Molecular Formula: C27H30O5S Formula Weight: 466.60 Physical Form: Crystalline Melting Point: 223° dec. Solubility: Insoluble in water. Soluble in alcohol, dilute alkali solutions MDL No.: MFCD00005869
pH indicator: 1.2 (red) to 2.8 (yellow) to 9.6 (blue)
Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-61-9
- Formula:C₂₇H₃₀O₅S
- MDL Number:MFCD00005869
- Molecular Weight:466.6 g/mol
- Merck Index:14,9400
- Melting Pt:223 °C (dec.)
- Storage Temperature:Ambiente
- EINECS:200-973-3
Documents
Specifications
Specifications
- Tipo di conf.:Bottiglia di plastica
- Conf:2 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



