EDTA ≥99.4% ACS
Bottiglia di vetro, 500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99.4+% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Physical Form: Powder Appearance: White Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541
Chelating agent
Synonyme: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- CAS Nummer:60-00-4
- Formel:C₁₀H₁₆N₂O₈
- MDL-Nummer:MFCD00003541
- Molekulargewicht:292.25 g/mol
- Merck-Index:14,3517
- Siedepunkt:614 °C (1013 hPa)
- Schmelzender Platingehalt:∼ 245 °C (dec.)
- Dichte:0,86 g/cm³ (20 °C)
- Blitz Pt:>100 °C
- Lagertemperatur:Ambiente
- EINECS:200-449-4
- ERREICHEN:01-2119486399-18
Dokumente
Technische Daten
Technische Daten
- Tipo di conf.:Bottiglia di vetro
- Conf:500 g



