Pycnometers, Hubbard
Pycnometer, conical, unadjustable, 50 ml, 24/15
Codice catalogo NEUB0303-50-50
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Vai a
Product Details & Documents
Borosilicate glass 3,3.
- With glass stopper
- For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses
According to DIN 12806 and DIN ISO 3507.
Specifiche
- Cono (NS):24/15
- Descrizione:Pycnometer, conical, unadjustable, 50 ml
- Seq:03