Orders LinkContactUsLinkComponent
Pycnometers, Hubbard
Pycnometer, conical, unadjustable, 25 ml, 24/15
  NEUB0303-50-25
Pycnometers, Hubbard
  NEUB0303-50-25
Pycnometers, Hubbard
Pycnometer, conical, unadjustable, 25 ml, 24/15
SFr. 133.00
1 * 1 ST
Borosilicate glass 3,3.
  • With glass stopper
  • For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses
According to DIN 12806 and DIN ISO 3507.
  • Cono (NS):
    24/15
  • Descrizione:
    Pycnometer, conical, unadjustable, 25 ml
  • Seq:
    02