Orders LinkContactUsLinkComponent
Pycnometers, Hubbard
Pycnometer, cylindrical, unadjustable, 24 ml, 24/15
  NEUB0303-50-24
Pycnometers, Hubbard
  NEUB0303-50-24
Pycnometers, Hubbard
Pycnometer, cylindrical, unadjustable, 24 ml, 24/15
SFr. 110.00
1 * 1 ST
Borosilicate glass 3,3.
  • With glass stopper
  • For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses
According to DIN 12806 and DIN ISO 3507.
  • Cono (NS):
    24/15
  • Descrizione:
    Pycnometer, cylindrical, unadjustable, 24 ml
  • Seq:
    01