Multiple or single cuvette segments
Cuvettes in pre-racked segments (15), PMMA
Codice catalogo KART1500
Nome del fornitore: KARTELL
Vai a
Product Details & Documents
These twelve position cuvettes segments have been specially designed for use with the Cobas Mira® analyser.
Confezione: In cases of bulk segments (450) or 30 pre-racked segments (15).
Specifiche
- Descrizione:Cuvettes in pre-racked segments (15)
- Materiale:PMMA
- Seq:002