Container, Securitainer®
Securitainer®-cap, 26 mm
Codice catalogo COVEC26/01
Nome del fornitore: COVERA
Vai a
Product Details & Documents
SECURITAINER®, made of white PP is an effective security container. It has two main uses: as an individual transport container for glass vials and as packaging for materials and substances sensitive to dampness.
- The tamper-proof seal guarantees that the packaging has not been opened-providing increased product security
- The seal prevents manipulation of vials, labels and their contents
- The strong, light construction protects the contents effectively
- Easy to handle
- Lids to be ordered seperately
Specifiche
- Dimensione tappo:26 mm
- Descrizione:Securitainer®-cap
- Seq:11