Wire baskets, Remanit® 4301
200×400×200 mm
Codice catalogo RSGA120.040
Nome del fornitore: RSG
Vai a
Product Details & Documents
These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
- Very good temperature resistance
- Good chemical resistance
- Reinforced edges
Specifiche
- L×P×A:200×400×200 mm