Orders LinkContactUsLinkComponent
Wire baskets, Remanit® 4301
Wire baskets, Remanit® 4301
200×300×200 mm
Codice catalogo RSGA120.030
Nome del fornitore:   RSG
Wire baskets, Remanit® 4301
Codice catalogo RSGA120.030
Nome del fornitore:  RSG
Wire baskets, Remanit® 4301
Wire baskets, Remanit® 4301
200×300×200 mm
Price
195.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
  • Very good temperature resistance
  • Good chemical resistance
  • Reinforced edges
Specifiche
  • L×P×A:
    200×300×200 mm