Wire baskets, Remanit® 4301
180×180×180 mm
Catalog # RSGA120.018
Supplier: RSG
Jump to:
Product Details & Documents
These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
- Very good temperature resistance
- Good chemical resistance
- Reinforced edges
Specifications
- L×P×A:180×180×180 mm