Orders LinkContactUsLinkComponent
Wire baskets, Remanit® 4301
180×180×180 mm
Catalog # RSGA120.018
Supplier:   RSG
Wire baskets, Remanit® 4301
Catalog # RSGA120.018
Supplier:  RSG
Wire baskets, Remanit® 4301
Wire baskets, Remanit® 4301
180×180×180 mm
Price
SFr. 144.00
Log in for availability
1 * 1 ST
Product Details & Documents
These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
  • Very good temperature resistance
  • Good chemical resistance
  • Reinforced edges
Specifications
  • L×P×A:
    180×180×180 mm