Orders LinkContactUsLinkComponent
Wire baskets, Remanit® 4301
Wire baskets, Remanit® 4301
160×160×160 mm
Codice catalogo RSGA120.016
Nome del fornitore:   RSG
Wire baskets, Remanit® 4301
Codice catalogo RSGA120.016
Nome del fornitore:  RSG
Wire baskets, Remanit® 4301
Wire baskets, Remanit® 4301
160×160×160 mm
Price
SFr. 139.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
  • Very good temperature resistance
  • Good chemical resistance
  • Reinforced edges
Specifiche
  • L×P×A:
    160×160×160 mm