Orders LinkContactUsLinkComponent
Wire baskets, Remanit® 4301
140×140×140 mm
  RSGA120.014
 :   RSG
Wire baskets, Remanit® 4301
  RSGA120.014
 :  RSG
Wire baskets, Remanit® 4301
Wire baskets, Remanit® 4301
140×140×140 mm
SFr. 133.00
1 * 1 ST
These electropolished wire baskets are made of stainless steel ¹⁸/₁₀.
  • Very good temperature resistance
  • Good chemical resistance
  • Reinforced edges
  • L×P×A:
    140×140×140 mm