Measuring beakers with handle
1000 ml, 130 mm, 100 mm
Codice catalogo 213-1037
Nome del fornitore: RSG
Vai a
Product Details & Documents
Stainless steel, rust-proof, with flared rim, spout and handle.
- Good chemical resistance
- Excellent resistance to heat
- Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Specifiche
- Ø est:100 mm
- Capacità:1000 ml
- Altezza:130 mm
- Seq:04