VALVE PTFE ROTARY 3-WAY(1.5MM BORE) 1 * 1 pezzi
Référence 554-2018
Fournisseur: IDEX RHEODYNE
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Fournisseur: VWR Chemicals
VWR®, Guanti in Nitrile
Fournisseur: VWR Collection
Strisce reattive pH, MQuant®, Supelco®
Fournisseur: Merck
VWR®, Salviette per il viso
Fournisseur: VWR Collection
Acqua, HiPerSolv CHROMANORM® per HPLC
Fournisseur: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Fournisseur: VWR Chemicals
Sealing film for manual application, Parafilm® M
Fournisseur: HEATHROW
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
VWR® NITRILE LIGHT, Guanti in nitrile
Fournisseur: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
Fournisseur: VWR Collection
VWR®, Pipette sierologiche, Standard Line
Fournisseur: VWR Collection
Guanti in nitrile, Kimtech™ Purple Nitrile™
Fournisseur: KIMBERLY CLARK