Orders LinkContactUsLinkComponent
Small flanges KF, screw-on
Small flanges KF, screw-on, internal thread, DN 25, Acciaio inossidabile
  VACU20672002
 :   VACUUBRAND
Small flanges KF, screw-on
  VACU20672002
Small flanges KF, screw-on
Small flanges KF, screw-on, internal thread, DN 25, Acciaio inossidabile
183.00 SFr.
1 * 1 ST
Small flanges made of stainless steel with sealing ring made of NBR.
Used for vacuum system.
  • Descrizione:
    Small flanges KF, screw-on, internal thread, DN 25
  • Materiale:
    Acciaio inossidabile
  • Seq:
    03