Small flanges KF, screw-on
Small flanges KF, screw-on, internal thread, DN 25, Acciaio inossidabile
VACU20672002
Small flanges made of stainless steel with sealing ring made of NBR.
Used for vacuum system.
- Descrizione:Small flanges KF, screw-on, internal thread, DN 25
- Materiale:Acciaio inossidabile
- Seq:03