Small flanges KF, screw-on
Small flanges KF, screw-on, internal thread, DN 16, Acciaio inossidabile
Katalognr. 185-2120
Anbieter: VACUUBRAND
Direkt zu
Product Details & Documents
Small flanges made of stainless steel with sealing ring made of NBR.
Used for vacuum system.
Dokumente
MB_KFAnschraubflansche
Technische Zeichnung
Technische Daten
- Descrizione:Small flanges KF, screw-on, internal thread, DN 16
- Materiale:Acciaio inossidabile
- Seq:02