Orders LinkContactUsLinkComponent
Small flanges KF, screw-on | Avantor
Small flanges KF, screw-on
Small flanges KF, screw-on, internal thread, DN 16, Acciaio inossidabile
Katalognr. 185-2120
Anbieter:   VACUUBRAND
Small flanges KF, screw-on
Katalognr. 185-2120
Anbieter:  VACUUBRAND
Small flanges KF, screw-on
Small flanges KF, screw-on, internal thread, DN 16, Acciaio inossidabile
Preis
SFr. 158.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Small flanges made of stainless steel with sealing ring made of NBR.
Used for vacuum system.
Dokumente
Technische Daten
  • Descrizione:
    Small flanges KF, screw-on, internal thread, DN 16
  • Materiale:
    Acciaio inossidabile
  • Seq:
    02