Reducing cross pieces with small flanges
Cross Piece, DN 25/25/16/16
Codice catalogo 185-2471
Nome del fornitore: VACUUBRAND
Vai a
Product Details & Documents
Reducing cross pieces with small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Documents
MB_Messkreuzstuecke_KF
Disegno tecnico
Specifiche
- Descrizione:Cross Piece, DN 25/25/16/16
- Seq:01