Mohr pinch cocks
80 mm
Codice catalogo 229-0335
Nome del fornitore: USBECK Laborgeräte
Vai a
Product Details & Documents
Nickel-plated brass. For regulating or stopping the flow of liquids in tube connections.
- Very good thermal resistance
- Easy handling
- Easy cleaning
Specifiche
- Lunghezza:80 mm