Tweezers
Blunt, 130 mm
Codice catalogo 232-0088
Nome del fornitore: USBECK Laborgeräte
Vai a
Product Details & Documents
Stainless steel. A large range of quality forceps for various laboratory applications, production applications etc.
- Practical
- Anti-magnetic
- Resistant to corrosives
Documents
Specifiche
- Tipo:Blunt
- Lunghezza:130 mm