Closures for headspace vials
PTFE coated silicone cap/spring, unassembled
Codice catalogo PERCB0104242
Nome del fornitore: PerkinElmer
Vai a
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifiche
- Descrizione:PTFE coated silicone cap/spring, unassembled
- Seq:03