Post-mortem scalpel handles
PM40 stainless steel, Blade no's PM40 and PM40B, 220 mm
Anbieter: SWANN MORTON
Direkt zu
Product Details & Documents
A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
- PM40 range with traditional screw fitting
- PM60 range with standard blade fitting
PM40B and PM60B blades are 'bull nose' blades for added safety and protection against accidental sharps injuries.
Verpackung: The PM8 and PM40 stainless steel handles and all blades are supplied individually wrapped.
Technische Daten
- Per:Blade no's PM40 and PM40B
- Tipo:PM40 stainless steel
- Lunghezza:220 mm
- Seq:02