pH indicator strips, Whatman™
Cartine indicatrici universali, strisce non sanguinanti, 0 - 14,0
Nome del fornitore: Whatman products (Cytiva)
pH indicator strips, Whatman™
Codice catalogo 700-0442
Nome del fornitore: Whatman products (Cytiva)
Vai a
Product Details & Documents
Universal indicator strips which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Convenient and portable for field use
Specifiche
- Campo pH:0 - 14,0
- Descrizione:Cartine indicatrici universali, strisce non sanguinanti