Transfer tubes, Transfertube®
Tubi di trasferimento, PP, 4 ml, 175 mm
Codice catalogo SPEC190195P
Nome del fornitore: Spectrum Laboratories
Vai a
Product Details & Documents
Disposable tubes for liquid and solid samples with one hand. The sample to be transfered is inserted with harvester and discharged using the plunger.
- Quick and easy use with the finger ring
- Reduces the risk of cross contamination
Recommended for particulates, suspensions, and microbial colonies.
Specifiche
- Capacità:4 ml
- Descrizione:Tubi di trasferimento
- Lunghezza:175 mm
- Materiale:PP
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



