Orders LinkContactUsLinkComponent
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography anion multi-element standard solution II, (3 elements in water) 1000 mg/l: Cl, NO3, SO4 Certipur®, 500 mL
Codice catalogo 1.11448.0500
Nome del fornitore:   Merck
Ion chromatography Multi-Element standards, Supelco®
Codice catalogo 1.11448.0500
Nome del fornitore:  Merck
Numero CAS:  
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography anion multi-element standard solution II, (3 elements in water) 1000 mg/l: Cl, NO3, SO4 Certipur®, 500 mL
Numero CAS:  
Price
SFr. 136.00
Log in per disponibilità
1 * 500 mL
Product Details & Documents
All ion chromatography standards are traceable to NIST standard reference materials.
Specifiche
Specifiche
  • Conf:
    500 mL
  • Descrizione:
    Ion chromatography anion multi-element standard solution II, (3 elements in water) 1000 mg/l: Cl, NO<sub>3</sub>, SO<sub>4 </sub>Certipur®
  • Cat. No.:
    1.11448.0500
  • M-Clarity quality level:
    MQ100