ICP multi-element standards, CertiPUR®, Supelco®
ICP multi-element standard solution VI, for ICP-MS (30 elements in dilute nitric acid) CertiPUR®, Ag, Al, Ba, Bi, Cd, Co, Cr, Cu, Ga, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Rb, Sr, Te, Tl, U, V: 10 mg/l; As, B, Be, Fe, Se, Zn: 100 mg/l; Ca 1000 mg/l, 6% HNO₃, bottiglia in plastica, 100 mL
Nome del fornitore: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Codice catalogo 1.10580.0100
Nome del fornitore: Merck
Numero CAS:
Vai a
Product Details & Documents
Safety note: These solutions are generally in acid solution.
Documents
Specifiche
Specifiche
- Matrice:6% HNO₃
- Tipo di conf.:bottiglia in plastica
- Conf:100 mL
- Descrizione:ICP multi-element standard solution VI, for ICP-MS (30 elements in dilute nitric acid) CertiPUR®
- Elements and Concentration:Ag, Al, Ba, Bi, Cd, Co, Cr, Cu, Ga, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Rb, Sr, Te, Tl, U, V: 10 mg/l; As, B, Be, Fe, Se, Zn: 100 mg/l; Ca 1000 mg/l
- Cat. No.:1.10580.0100
- M-Clarity quality level:MQ100
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



