Ion chromatography Multi-Element standards, Supelco®
Ion chromatography cation multi-element standard solution I, (9 elements in diluted nitic acid) 100 mg/l: Li, Na, K, NH4, Mn, Ca, Mg, Sr, Ba Certipur®, 100 mL
Nome del fornitore: Merck
Ion chromatography Multi-Element standards, Supelco®
Codice catalogo 1.10322.0100
Nome del fornitore: Merck
Numero CAS:
Vai a
Product Details & Documents
All ion chromatography standards are traceable to NIST standard reference materials.
Documents
Certificates
Cerca per numero di lottoCertipur
Letteratura sul prodotto
msds_109032 msds_109036 msds_110322 msds_111437 msds_111448
Scheda di sicurezza
More
Specifiche
Specifiche
- Conf:100 mL
- Descrizione:Ion chromatography cation multi-element standard solution I, (9 elements in diluted nitic acid) 100 mg/l: Li, Na, K, NH<sub>4</sub>, Mn, Ca, Mg, Sr, Ba<sub> </sub>Certipur®
- Cat. No.:1.10322.0100
- M-Clarity quality level:MQ100