GC capillary column tools
Replacement PTFE 1/16" sealing ring
Codice catalogo 558-0190
Nome del fornitore: Trajan Scientific and Medical
GC capillary column tools
Codice catalogo 558-0190
Nome del fornitore: Trajan Scientific and Medical
Vai a
Product Details & Documents
The capillary cutting tool is used for burr free cutting of fused silica tubing.
- Reduces likelihood of blockages
- Ensures square ends for 'Press fit' style connectors
- Clean entry for on-column injection
Specifiche
- Descrizione:Replacement PTFE 1/16" sealing ring
- Seq:02
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.