Syringes with valves
Syringe pre-fitted with Luer lock valve, PTFE tipped plunger, 50 ml, 50MR-VLLMA-GT
Codice catalogo SGEA009670
Nome del fornitore: Trajan Scientific and Medical
Vai a
Product Details & Documents
These syringes are pre-fitted with valves. They are ideal for sample storage and transportation or to pressurise samples prior to injection.
Documents
Specifiche
- Tipo:50MR-VLLMA-GT
- Volume:50 ml
- Descrizione:Syringe pre-fitted with Luer lock valve, PTFE tipped plunger
- Seq:11