Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Ion exchangers

Ion exchangers

Strong basic anion exchanger, Ion exchanger III, OH-, exchange capacity >0,9 mval/ml, 500 g
Codice catalogo 1.04767.0500
Nome del fornitore:   Merck
Ion exchangers
Codice catalogo 1.04767.0500
Nome del fornitore:  Merck
Numero CAS:  
Ion exchangers

Strong basic anion exchanger, Ion exchanger III, OH-, exchange capacity >0,9 mval/ml, 500 g

Numero CAS:  
Price
SFr. 376.00
Log in per disponibilità
1 * 500 g
Product Details & Documents

In general, ion exchangers consist of a cross-linked polymer matrix with a uniform distribution of fixed ionic groups sited throughout the resin structure. These must be balanced by a similar number of ions of the opposite charge, (counter ions) to maintain electrical neutrality. Cation exchangers therefore, exchange and enrich only cations, anion exchangers only anions. In contrast adsorber resins have a non ionic, but depending on the structure, a somewhat polar character, and don't adsorb stoichiometrically both: anions, cations, as well as uncharged compounds.

Possible applications are:
Trace enrichment by using chelating ion exchangers
Determination of total salt content of solutions and water by H+ exchange
Removals of interfering cations or anions
Chromatographic separation
Digestion of insoluble compounds
Application as a catalyst

Specifiche

Specifiche

  • Conf:
    500 g
  • Descrizione:
    Strong basic anion exchanger, Ion exchanger III, OH<sup>-</sup>, exchange capacity >0,9 mval/ml

Specifiche