Stopcocks, for storage tanks, Nalgene®
12,7 mm NPT, male thread
Codice catalogo NALG6420-0500
Nome del fornitore: Thermo Fisher Scientific
Stopcocks, for storage tanks, Nalgene®
Codice catalogo NALG6420-0500
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
LDPE, white.
- Good chemical resistance
- Godd temperature resistance
Ensure safe and efficient liquid dispensing from tanks by using Nalgene™ spigots for storage tanks. Spigots are for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.
Specifiche
- Filettatura:12,7 mm NPT, male thread
- Cat. No.:NALG6420-0500
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:NAR Certificate of Retirement