Giare con coperchio, Nalgene®
2200 ml, 132 mm, 193 mm
Katalognr. NALG5352-0002
Anbieter: Thermo Fisher Scientific
Giare con coperchio, Nalgene®
Katalognr. NALG5352-0002
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
PP, translucent.
- Straight sides and no neck making access and retrieval easy
- Larger sizes make excellent waste crocks
- Autoclavable
Ideal for storage and transport of solids.
Technische Daten
- Ø est:132 mm
- Capacità:2200 ml
- Altezza:193 mm
- Cat. No.:NALG5352-0002
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)