Pipeline filters, DURAN®
Pipeline filter, 40 - 100 µm, 90 mm
Catalog # 511-0422
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These in-line filters are used for in-line filtration of gas lines to remove solid impurities (e.g. dust).
Made from DURAN®, borosilicate glass 3,3, clear.
Specifications
- Ø:90 mm
- Dimensioni pori:40 - 100 µm
- Descrizione:Pipeline filter