Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Sequagel Buffer 1 * 1 l

Sequagel Buffer 1 * 1 l

Codice catalogo NADIEC-840-1L
Marketsource
Nome del fornitore:   NATIONAL DIAGNOSTICS
Sequagel Buffer 1 * 1 l
Codice catalogo NADIEC-840-1L
Nome del fornitore:  NATIONAL DIAGNOSTICS
Marketsource
Sequagel Buffer 1 * 1 l
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 L
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.