Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Becher, Philips, DURAN®

Becher, Philips, DURAN®

500 ml, 142 mm, 86 mm
Catalog # 213-0215
Supplier:   DWK Life Sciences
Becher, Philips, DURAN®
Catalog # 213-0215
Supplier:  DWK Life Sciences
Becher, Philips, DURAN®
Becher, Philips, DURAN®

500 ml, 142 mm, 86 mm

Price
SFr. 29.70
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, vetro borosilicato 3.3 trasparente, con beccuccio.

  • Ottima resistenza chimica
  • Elevata resistenza termica
  • Ridotta dilatazione termica

Pareti spesse con graduazioni stampate e area di scrittura.

Specifications
  • Ø est:
    86 mm
  • Capacità:
    500 ml
  • Altezza:
    142 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.