Orders LinkContactUsLinkComponent
Adapters, delivery, bent
Adapters, delivery, bent
14/23 NS, 14/23 NS
Codice catalogo LENZ05265001
Nome del fornitore:   Lenz Laborglas
Adapters, delivery, bent
Codice catalogo LENZ05265001
Nome del fornitore:  Lenz Laborglas
Adapters, delivery, bent
Adapters, delivery, bent
14/23 NS, 14/23 NS
Price
SFr. 31.20
Log in per disponibilità
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.
Documents
Specifiche
  • Dimensione cono:
    14/23 NS
  • Cono femmina:
    14/23 NS