Distillatori
250 mm, 29/32 NS, 14/23 NS
Codice catalogo 201-3131
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Bridge includes fused Liebig condenser and curved vacuum adapter. NS 14/23 socket for distillation thermometer with 55 mm fitted length. Water and vacuum connection via hose connectors with 8 mm external diameter or water connection with thread GL 14 and vacuum receiver adapter with thread GL 18.
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Schott tubing
Scheda dati
Specifiche
- Dimensione cono:29/32 NS
- Lunghezza mantello:250 mm
- Cono femmina:14/23 NS
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



