Imbuti Büchner
6000 ml, 240 mm
Codice catalogo 519-2367
Nome del fornitore: KARTELL
Vai a
Product Details & Documents
PP, white.
- Robust and lightweight
- Top and bottom can be separated for easy cleaning
For filter paper or membrane filtration discs.
Specifiche
- Capacità:6000 ml
- per Ø filtro:240 mm