Rubinetto per tubi
4,9 mm, 8 mm
Catalog # 227-4008
Supplier: KARTELL
Jump to:
Product Details & Documents
HDPE con maschio in PP
- Good chemical resistance
- No lubricants required
- Good thermal resistance
These flow through valves are ideal for regulating or stopping the flow of gases and liquids in tube connections under low pressure (up to 0,5 bar) or partial vacuum. Smooth connectors, slightly tapered on both sides.
Specifications
- Ø foro:4,9 mm
- Seq:01
- per tubo con Ø esterno:8 mm