Fumaroyl chloride 95%
Bottiglia di vetro, 500 mL
Supplier: Thermo Fisher Scientific
Fumaroyl chloride 95%
Catalog # ACRO165975000
Supplier: Thermo Fisher Scientific
CAS Number: 627-63-4
Jump to:
Product Details & Documents
Synonyms: Fumaryl chloride
- CAS Number:627-63-4
- Formula:C₄H₂Cl₂O₂
- MDL Number:MFCD00000733
- Molecular Weight:152.96 g/mol
- Boiling Pt:161…164 °C (760 mmHg)
- Melting Pt:–2 °C
- Density:1,41 g/cm³
- Flash Pt:73 °C
- Storage Temperature:Refrigerata
- EINECS:211-005-4
- UN:1780
- ADR:8,II
Documents
ACRO16597
SDS
Specifications
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4980 to 1.5010 (20°C, 589 nm)
- GC:≥94 %
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



