Bottle pourers
Bottle pourer, PTFE, blue, for GL 32 screw neck bottles
Codice catalogo COWI028.032
Nome del fornitore: COWIE
Vai a
Product Details & Documents
Manufactured in pure, FDA approved, PTFE.
- Totally chemically resistant and inert
- To fit GL45, 38, 32 and 30 screw neck bottles
- State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.
Specifiche
- Tipo:for GL 32 screw neck bottles
- Descrizione:Bottle pourer, PTFE, blue